missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P2X2 Rabbit anti-Guinea Pig, Human, Mouse, Non-human primate, Porcine, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA124624
Additional Details : Weight : 0.01000kg
Description
P2rx2 are members of the ligand-gated ion channel family that open in response to extracellular ATP. Each receptor is made up of a trimer of subunits (P2X1-7) all of which share the common structure of two transmembrane domains. P2rx2 functions as a ligand-gated ion channel and binding to ATP mediates synaptic transmission between neurons and from neurons to smooth muscle. Six transcript variants encoding six distinct isoforms have been identified for P2rx2. P2rx2 is involved in a variety of cellular responses, such as excitatory postsynaptic responses in sensory neurons, neuromuscular junctions (NMJ) formation, hearing, perception of taste and peristalsis. In the inner ear, P2rx2 regulates sound transduction and auditory neurotransmission, outer hair cell electromotility, inner ear gap junctions, and K(+) recycling. P2rx2 also mediates synaptic transmission between neurons and from neurons to smooth muscle. Diseases associated with P2RX2 include Deafness, Autosomal Domit 41 and Dfna41 Nonsyndromic Hearing Loss And Deafness.Specifications
| P2X2 | |
| Polyclonal | |
| whole serum with 0.05% sodium azide | |
| O70397, P51575, Q9UBL9, P51576, Q8K3P1, P47824, P49653 | |
| MGC129601, P2X2 | |
| Rabbit | |
| IgG | |
| RUO | |
| Antibody | |
| Polyclonal | |
| Guinea Pig, Human, Mouse, Non-human primate, Porcine, Rat |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| P2RX1 | |
| Liquid | |
| P2RX1, P2RX2 | |
| Synthetic peptide corresponding to residues D(460) S T S T D P K G L A Q L(472) of rat P2X2. | |
| 50 μL | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Primary | |
| 100135591, 100727772, 100736581, 106504105, 114115, 18436, 22953, 231602, 25505, 5023 |