missing translation for 'onlineSavingsMsg'
Learn More

beta Actin Rabbit anti-Avian, Fish, Hamster, Human, Mouse, Non-human primate, Porcine, Rabbit, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA146296

Additional Details : Weight : 0.01000kg

Product Code. 15618038

  • 390.00€ / 50µL


EU Stock 4
Add to basket

Description

Description

Beta actin is one of six actin isoforms which have been identified. Beta actin is a non-muscle cytoskeletal protein in all human cell types and is involved in cell motility, structure, and integrity. There are six different actin isoforms in human. The beta isoform of actin, along with gamma actin, coexist in most cell types as components of the cyto-skeleton. Beta actins are cytoplasmic proteins ubuquitously expressed in all eukaryotic cells. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of two stranded helix. Actins are highly conserved proteins that are involved in cell motility, structure and integrity. Because beta actin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting. Antibodies to beta-actin provide a specific and useful tool in studying the intracellular distribution of beta-actin and the static and dynamic aspects of the cytoskeleton.
Specifications

Specifications

beta Actin
Polyclonal
Unconjugated
ACTB
Liquid
ACTB
Synthetic peptide derived from the N-terminus of Human beta-Actin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Avian, Fish, Hamster, Human, Mouse, Non-human primate, Porcine, Rabbit, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
1 mg/mL
tris citrate, tris phosphate with 0.09% sodium azide; pH 7-8
P60709, P60710, Q6QAQ1, P29751, P60711
PS1TP5BP1, Atcb
Rabbit
IgG
50 μL
4° C, do not freeze
Primary
100009272, 100689477, 11461, 414396, 60, 81822
Videos