missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6x-His Tag Goat anti-Tag, HRP, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA123024
Additional Details : Weight : 0.01000kg
Description
This antibody does not detect all N-terminal 6-His tagged proteins.Antibody concentration was determined by extinction coefficient prior to conjugation: absorbance at280 nm of 1.4 equals 1.0 mg of IgG. Molar enzyme/antibody protein ratio is 4:1.Specifications
| 6x-His Tag | |
| Polyclonal | |
| HRP | |
| Liquid | |
| Goat | |
| IgG | |
| 100 μg | |
| 4° C, do not freeze | |
| Primary | |
| Tag |
| ELISA, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| 50mM HEPES buffered saline with 0.1M NaCl, 0.2% BSA and 0.01% thimerosal; pH 7.1 | |
| His Tag; Histidine Tag; Histidine Epitope Tag; 6X His eptope tag; 6His tag; 6X His | |
| 6-His (HHHHHH) conjugated to KLH | |
| Antigen affinity chromatography | |
| RUO | |
| Antibody | |
| Polyclonal |