Learn More
Eppendorf™ Aluminum Fixed Angle Rotors
Use these rotors for a variety of applications. Eppendorf™ Aluminum Fixed Angle Rotors are ideal for molecular and biological laboratories.
545.00€ - 2625.00€
Product Code | Brand | Capacity | Includes | Max. Speed | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Includes | Max. Speed | Price | Quantity & Availability | |||
10422662
|
Eppendorf™
5424706002 |
18 x 1.5/2 mL | Polypropylene Lid | 14,680 rpm |
605.00€
Each |
In Stock
Log in to see stock. |
|||
10555563
|
Eppendorf™
5424702007 |
24 x 1.5/2 mL | Aerosol rotor lid | 14,680 rpm |
765.00€
Each |
In Stock
Log in to see stock. |
|||
10634603
|
Eppendorf™
5424704000 |
4 x 0.2 mL | Aerosol rotor lid | 14,680 rpm |
545.00€
Each |
In Stock
Log in to see stock. |
|||
10733912
|
Eppendorf™
5820715006 |
6 x 15/50 mL | 12,100 rpm |
2625.00€
Each |
In Stock
Log in to see stock. |
||||
10368522
|
Eppendorf™
5427716009 |
6 x 15/50 mL | Screw-on lid | 7,830 rpm |
1465.00€
Each |
In Stock
Log in to see stock. |
|||
10665753
|
Eppendorf™
5427714006 |
64 x 0.2 or 8 x 8 mL | Screw-on lid | 11,800 rpm |
895.00€
Each |
In Stock
Log in to see stock. |
|||
Description
- Choice of four different rotors
- Standard rotor can accommodate 1.5mL/2.0mL tubes
- PCR* strips and kits/spin columns can be centrifuged in the corresponding rotor
Specifications
Aluminum | |
Fixed Angle Rotor |