missing translation for 'onlineSavingsMsg'
Learn More

ALDH3A1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA584050

Additional Details : Weight : 0.01000kg

Product Code. 16331005

  • 530.00€ / 100µL
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

Immunogen sequence: NSGMGSYHGK KSFETFSHRR SCLVRPLMND EGLKVRYPPS PAKMTQH

Aldh3A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldh3A1 is highly expressed in stomach and even more strongly in cornea, representing between 5 to 50% of the water soluble protein fraction in mammalian corneas. It is thought that Aldh3A1 acts to protect the cornea from UV-induced oxidative stress by not only detoxification of reactive aldehydes by also through the direct absorption of UV energy. However, corneas from Aldh3A1-null mice are indistinguishable from those from wild-type mice; mice lacking both Aldh3A1 and Aldh1A1 showed increased cataract formation following UVB exposure, suggesting that Aldh1A1 may be able to compensate for the loss of Aldh3A1.
Specifications

Specifications

ALDH3A1
Polyclonal
Unconjugated
ALDH3A1
Ahd4, Ahd-4, AHDC, Ahd-c, alcohol dehydrogenase family 3, subfamily A1, Aldd, Aldehyde dehydrogenase (Ahd-c), Aldehyde dehydrogenase 3, aldehyde dehydrogenase 3 family member A1, aldehyde dehydrogenase 3 family, member A1, aldehyde dehydrogenase 3 family, memberA1, aldehyde dehydrogenase 3, stomach cytosolic (class 3), aldehyde dehydrogenase 3A1, aldehyde dehydrogenase 4, aldehyde dehydrogenase class 3, aldehyde dehydrogenase family 3 member A1, aldehyde dehydrogenase family 3 subfamily A1, aldehyde dehydrogenase family 3, member A1, aldehyde dehydrogenase family 3, subfamily A1, aldehyde dehydrogenase isozyme 3, aldehyde dehydrogenase type III, aldehyde dehydrogenase, dimeric NADP-preferring, Aldh, Aldh3, Aldh3a1, Aldh4, ALDHIII, BCP54, bovine corneal protein 54, corneal 15.8 kDa protein, corneal epithelium, corneal protein 54, transparentin, dehydrogenase 3, dimeric NAPD-prefering, dioxin-inducible aldehyde dehydrogenase 3, HTC-ALDH, MGC10406, stomach aldehyde dehydrogenase, transpar
Rabbit
Anitgen affinity chromatography, Antigen affinity chromatography
RUO
218
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry, Western Blot, Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide, pH 7.2
P30838
ALDH3A1
Recombinant protein corresponding to Human ALDH3A1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers