Learn More
Thermo Scientific™ Adapters for HIGHConic III Rotor
Assorted adapters to be used with the HIGHConic III rotor
135.00€ - 293.00€
Product Code | Brand | Capacity | Max. Tube Diameter | Max. Tube Length | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Max. Tube Diameter | Max. Tube Length | Price | Quantity & Availability | |||
15798139
|
Thermo Scientific™
75005802 |
38 mL | 25.5 mm | 110 mm |
167.00€
Pack of 2 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
15708149
|
Thermo Scientific™
75005803 |
16 mL | 18 mm | 123 mm |
167.00€
Pack of 2 |
In Stock
Log in to see stock. |
|||
15738578
|
Thermo Scientific™
75005804 |
12 mL | 16 mm | 96 mm |
167.00€
Pack of 2 |
In Stock
Log in to see stock. |
|||
15718149
|
Thermo Scientific™
75005805 |
6.5 mL | 13.5 mm | 114 mm |
167.00€
Pack of 2 |
In Stock
Log in to see stock. |
|||
15728149
|
Thermo Scientific™
75005806 |
3.5 mL | 11 mm | 100 mm |
167.00€
Pack of 2 |
In Stock
Log in to see stock. |
|||
15753569
|
Thermo Scientific™
75005807 |
1.5/2.0 mL | 11 mm | 40 mm |
167.00€
Pack of 2 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
15105273
|
Thermo Scientific™
75005808 |
8 x 15 mL | 12 mm | 17 mm |
293.00€
1 set |
In Stock
Log in to see stock. |
|||
15271026
|
Thermo Scientific™
75005770 |
5 mL | 17 mm | 100 mm |
135.00€
Pack of 2 |
In Stock
Log in to see stock. |
|||
Description
Accommodate conical tubes in the 50mL buckets for use with the Thermo Scientific HIGHConic III fixed angle rotor.
- Adapters to run various tubes in HIGHConic III rotor
Specifications
Rotor Adapter |