Learn More
Abnova™ CCND1/CEN11q (Orange/Green) FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0113.200uL
Additional Details : Weight : 0.02000kg
Product Code. 16033175
  • 2430.00€ / 200µL
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
Store at -20°C in the dark.
Orange, Green
11q13/11p11.11-q11
CCND1/CEN11q FISH Probe
Genomic DNA
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Fluorescent In Situ Hybridization
Approx. 162kb/0.68kb
200 μL