Learn More
1KG AMMONIUM PEROXODISULFATE BAKER ANALYZED Reagent
Brand: Avantor Perfomance Materials 0762.1000
95.95 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
1KG AMMONIUM PEROXODISULFATE BAKER ANALYZED Reagent
Additional Details : CAS : 7727-54-0 Weight : 1.00000kg Transport : UN number : 1444 Chem class : 5.1 Pack group : III
Safety and Handling
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.