Learn More
Thermo Scientific™ 1-Step Human Coupled In Vitro Protein Expression Kits
Rapidly express full-length, functional proteins from mRNA or plasmid templates with yields of up to 100µg/mL per reaction using these cell-free kits.
87.50€ - 706.00€
| Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
| 10013544 |
Thermo Scientific™
88899 |
Pierce Recombinant GFP Protein | 20µg |
87.50€ 20µg |
|
12126510 |
Thermo Scientific™
88882 |
Pierce 1-Step Human Coupled IVT Kit - DNA | 40 rxn kit |
706.00€ 40 reactions |
from Supplier 30-04-2019 |
12116510 |
Thermo Scientific™
88881 |
Pierce 1-Step Human Coupled IVT Kit - DNA | 8 rxn kit |
186.00€ 8 reactions |
from Supplier 02-05-2019 |
15215743 |
Thermo Scientific™
88880 |
Pierce tGFP mRNA | 10μg |
127.00€ 10µg |
from Supplier 06-05-2019 |
Description
The Human IVT Kits are unique HeLa cell lysate-based protein expression systems for in vitro translation or coupled transcription/translation reactions. Protein expression is performed in a single 90-minute reaction that can be extended for up to 6 hours with continued protein production up to 100μg/mL when combined with the optimized pT7CFE1 Expression Vector. The Human IVT Kits can express functional enzymes, phosphoproteins, glycoproteins and membrane proteins for immediate use in studying protein interactions, performing rapid mutational analysis and measuring activity.
Highlights:
Functional – uses the human translational machinery to express active proteins
- Convenient – perform transcription and translation in a single step
- High performance – greater yields compared to rabbit reticulocyte in vitro translation
- Reliable – express proteins that fail in rabbit reticulocyte systems
Better Than Traditional Methods:
- HeLa cell-free extracts are capable of expressing proteins with post-translational modifications
- Accurate translation delivers full-length protein compatible with downstream applications
- Protein translation is optimized with EMCV IRES element and other mRNA stabilizing elements
Includes:
Complete kits contain HeLa cell lysate, accessory proteins, reaction mix, nuclease-free water, expression vector and a GFP-positive control vector
Requires:
30°C incubator or water bath
Recommended for:
Express proteins to measure enzyme activity; Perform mutational analysis; Express protein for use in gel mobility shift assays; Perform co-immunoprecipitation; Express cytotoxic proteins; Perform unnatural amino acid labeling