Learn More
Thermo Scientific™ 1-Step Human Coupled In Vitro Protein Expression Kits
Rapidly express full-length, functional proteins from mRNA or plasmid templates with yields of up to 100µg/mL per reaction using these cell-free kits.
87.50€ - 706.00€
Products 4
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
10013544 Thermo Scientific™
88899
Pierce Recombinant GFP Protein 20µg 87.50€
20µg View alternative products
Save Up to 
»
EU Stock 4
Add to basket
12126510 Thermo Scientific™
88882
Pierce 1-Step Human Coupled IVT Kit - DNA 40 rxn kit 706.00€
40 reactions View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 30-04-2019
Add to basket
12116510 Thermo Scientific™
88881
Pierce 1-Step Human Coupled IVT Kit - DNA 8 rxn kit 186.00€
8 reactions View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 02-05-2019
Add to basket
15215743 Thermo Scientific™
88880
Pierce tGFP mRNA 10μg 127.00€
10µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-05-2019
Add to basket
Description
Description
The Human IVT Kits are unique HeLa cell lysate-based protein expression systems for in vitro translation or coupled transcription/translation reactions. Protein expression is performed in a single 90-minute reaction that can be extended for up to 6 hours with continued protein production up to 100μg/mL when combined with the optimized pT7CFE1 Expression Vector. The Human IVT Kits can express functional enzymes, phosphoproteins, glycoproteins and membrane proteins for immediate use in studying protein interactions, performing rapid mutational analysis and measuring activity.
Highlights:
Functional – uses the human translational machinery to express active proteins
  • Convenient – perform transcription and translation in a single step
  • High performance – greater yields compared to rabbit reticulocyte in vitro translation
  • Reliable – express proteins that fail in rabbit reticulocyte systems
Better Than Traditional Methods:
  • HeLa cell-free extracts are capable of expressing proteins with post-translational modifications
  • Accurate translation delivers full-length protein compatible with downstream applications
  • Protein translation is optimized with EMCV IRES element and other mRNA stabilizing elements
Includes:
Complete kits contain HeLa cell lysate, accessory proteins, reaction mix, nuclease-free water, expression vector and a GFP-positive control vector
Requires:
30°C incubator or water bath
Recommended for:
Express proteins to measure enzyme activity; Perform mutational analysis; Express protein for use in gel mobility shift assays; Perform co-immunoprecipitation; Express cytotoxic proteins; Perform unnatural amino acid labeling